accessibility menu, dialog, popup

UserName

NaH3COHOO

CAS RN 5094-24-6

IUPAC Name: sodium 2-hydroxybutanoate
Synonyms: 2-hydroxybutyric acid
Molecular Weight (g/mol): 126.09
Molecular Formula: C4H7NaO3
InChi Key: MOSCXNXKSOHVSQ-UHFFFAOYNA-M
SMILES: [Na+].CCC(O)C([O-])=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)