missing translation for 'onlineSavingsMsg'
Learn More

Sodium Diphenylphosphinobenzene-3-sulfonate 90.0+%, TCI America™

Catalog No. D20435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20435G 5 g
D20431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20435G Supplier TCI America Supplier No. D20435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63995-75-5
Molecular Formula C18H14NaO3PS
Molecular Weight (g/mol) 364.33
MDL Number MFCD00144157
InChI Key NTUROZDXWLPVHB-UHFFFAOYSA-M
Synonym Diphenylphosphinobenzene-3-sulfonic Acid Sodium Salt, Triphenylphosphine-3-sulfonic Acid Sodium Salt
PubChem CID 23668331
IUPAC Name sodium 3-(diphenylphosphanyl)benzene-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=CC(=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 5 g
Formula Weight 364.33
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Diphenylphosphinobenzene-3-sulfonate