missing translation for 'onlineSavingsMsg'
Learn More

Sodium Diphenylamine-4-sulfonate 97.0+%, TCI America™

Supplier:  TCI America D08771G

 View more versions of this product

Catalog No. D08771G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Diphenylamine-4-sulfonate
Gray-White
MFCD00036468
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
272.27
271.27
Crystalline Powder
6152-67-6
C12H11NNaO3S
1 g
VMVSDVIYIMNZBS-UHFFFAOYSA-N
4-(phenylamino)benzene-1-sulfonic acid sodium
517002
≥97.0% (HPLC,N)

Chemical Identifiers

6152-67-6
272.27
VMVSDVIYIMNZBS-UHFFFAOYSA-N
517002
[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
C12H11NNaO3S
MFCD00036468
sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
4-(phenylamino)benzene-1-sulfonic acid sodium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions