missing translation for 'onlineSavingsMsg'
Learn More

Sodium Diphenylamine-4-sulfonate 97.0+%, TCI America™

Catalog No. p-7126480
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08771G 1 g
D087725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D08771G Supplier TCI America Supplier No. D08771G
Only null left

Chemical Identifiers

CAS 6152-67-6
Molecular Formula C12H11NNaO3S
Molecular Weight (g/mol) 272.27
MDL Number MFCD00036468
InChI Key VMVSDVIYIMNZBS-UHFFFAOYSA-N
Synonym sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
PubChem CID 517002
IUPAC Name 4-(phenylamino)benzene-1-sulfonic acid sodium
SMILES [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Color Gray-White
Quantity 1 g
Formula Weight 271.27
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Diphenylamine-4-sulfonate