Learn More
Sodium dihydrogen phosphate monohydrate, Puratronic™, 99.998% (metals basis)
CAS: 10049-21-5 | H5NaO5P | 139.00 g/mol
$287.74 - $884.42
Chemical Identifiers
| CAS | 10049-21-5 |
|---|---|
| Molecular Formula | H5NaO5P |
| Molecular Weight (g/mol) | 139.00 |
| MDL Number | MFCD00149208 |
| InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
| Synonym | sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 |
| PubChem CID | 516949 |
| IUPAC Name | sodium;dihydrogen phosphate;hydrate |
| SMILES | O.[Na+].OP(O)(O)=O |
Description
Sodium dihydrogen phosphate monohydrate is used as a food additive. It finds use as mordant in dyeing. It is also used in the manufacture of enamels, chemicals, pharmaceuticals, ceramics, detergents, and boiler compounds. In addition, it is used as a fireproofing agent, thickening agent, emulsifier, and as a reagent and buffer in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10049-21-5 | |
| 139.00 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| 516949 | |
| O.[Na+].OP(O)(O)=O |
| H5NaO5P | |
| MFCD00149208 | |
| sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
| sodium;dihydrogen phosphate;hydrate |
Specifications
| 100°C to H2O | |
| White | |
| 25 g | |
| H5NaO5P | |
| sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| sodium;dihydrogen phosphate;hydrate | |
| 516949 | |
| Odorless | |
| White powder | |
| Sodium dihydrogen phosphate monohydrate |
| 10049-21-5 | |
| Powder | |
| 99.998% (metals basis) | |
| MFCD00149208 | |
| Soluble in water; insoluble in ethanol,ether and chloroform. | |
| O.[Na+].OP(O)(O)=O | |
| 139.00 | |
| 137.99 (119.98 Anhydrous) | |
| Hygroscopic | |
| 2.04g/cm3 at 20°C |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber : 231-449-2
RTECSNumber : WA1900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only