Learn More
Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%
CAS: 10049-21-5 | H2NaO4P x H2O | 137.99 g/mol
Supplier: Thermo Scientific Chemicals 011591A1
Description
Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It is applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium dihydrogen phosphate monohydrate | |
| 10049-21-5 | |
| Crystalline | |
| 98.0-102.0% | |
| MFCD00149208 | |
| Highly soluble in water. Insoluble in ethanol,ether and chloroform. | |
| O.[Na+].OP(O)(O)=O | |
| 137.99 | |
| 137.99 (119.98 Anhydrous) | |
| Odorless | |
| White crystals |
| 100°C-H2O | |
| White | |
| 1 kg | |
| H2NaO4P x H2O | |
| sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| sodium phosphoric acid hydrate | |
| 516949 | |
| ACS | |
| Hygroscopic | |
| 2.04g/cm3 at 20°C |
Chemical Identifiers
| 10049-21-5 | |
| 137.99 | |
| BBMHARZCALWXSL-UHFFFAOYSA-N | |
| 516949 |
| H2NaO4P x H2O | |
| MFCD00149208 | |
| sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
| O.[Na+].OP(O)(O)=O |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber : 231-449-2
RTECSNumber : WA1900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only