missing translation for 'onlineSavingsMsg'
Learn More

Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%

Catalog No. AA1159130
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1159130 250 g
AA11591A1 1 kg
AA11591A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1159130 Supplier Thermo Scientific Chemicals Supplier No. 01159130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10049-21-5 ∣ H2NaO4P x H2O ∣ 137.99 g/mol

Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It is applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H2NaO4P x H2O
Molecular Weight (g/mol) 137.99
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O

Specifications

Melting Point 100°C (in 2H2O)
Physical Form Crystalline
Quantity 250 g
Odor Odorless
Sensitivity Hygroscopic
Merck Index 14,8660
Solubility Information Highly soluble in water. Insoluble in ethanol,ether and chloroform.
IUPAC Name sodium;dihydrogen phosphate;hydrate
Formula Weight 137.99 (119.98 Anhydrous)
Percent Purity 98.0 to 102.0%
Grade ACS
Chemical Name or Material Sodium dihydrogen phosphate monohydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 231-449-2
RTECSNumber WA1900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only