missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dihydrogen Phosphate Monohydrate

Catalog No. p-6970074
Change view
Click to view available options
Quantity:
500 gsol
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1063460500 500 gsol Plastic bottle
M1063461000 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1063460500 Supplier Technidata Supplier No. 1.06346.0500

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

pH 4.1 to 4.5 (50g/L, H2O, 25°C)
Quantity 500 gsol
Formula Weight 137.99densit
Percent Purity 99.0 to 102.0%
Grade ACS
Packaging Plastic bottle
Density 2.04 g/cm3 (20°C)
Chemical Name or Material Sodium dihydrogen phosphate monohydrate
EINECSNumber 231-449-2