missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dibenzyldithiocarbamate Hydrate 98.0+%, TCI America™

Catalog No. p-7126063
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01565G 5 g
D015625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D01565G Supplier TCI America Supplier No. D01565G
Only null left

Chemical Identifiers

CAS 55310-46-8
Molecular Formula C15H14NNaS2
Molecular Weight (g/mol) 295.394
MDL Number MFCD00066462
InChI Key MYCPTMLDBVIIES-UHFFFAOYSA-M
Synonym Dibenzyldithiocarbamic Acid Sodium Salt
PubChem CID 4676203
IUPAC Name sodium;N,N-dibenzylcarbamodithioate
SMILES C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 295.39
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dibenzyldithiocarbamate Hydrate