accessibility menu, dialog, popup

UserName

CAS RN 55310-46-8

NaSSN

CAS RN 55310-46-8

IUPAC Name: sodium (dibenzylcarbamothioyl)sulfanide
Synonyms: sodium (dibenzylcarbamothioyl)sulfanide
Molecular Weight (g/mol): 295.39
Molecular Formula: C15H14NNaS2
InChi Key: MYCPTMLDBVIIES-UHFFFAOYSA-M
SMILES: [Na+].[S-]C(=S)N(CC1=CC=CC=C1)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)