missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dehydroacetic Acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014208
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014208 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014208 Supplier Merck Emd Millipore Supplier No. PHR160910G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Sodium Dehydroacetic Acid belongs to the class of heterocyclic compounds, and has a variety of applications such as preservative in foods, cosmetics, pharmaceuticals, mold inhibitor in strawberries and others, antimicrobial, fungicide, plasticizer, etc.

Specifications

CAS 4418-26-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C8H7 NaO4
MDL Number MFCD00040583
Melting Point 295°C
Synonym Dehydroacetic acid sodium salt; 2-Acetyl-5-hydroxy-3-oxo-4-hexenoic acid delta-lactone sodium salt
Type Certified Reference Material
Quantity 10 g
Formula Weight 190.13
Grade Certified Reference Material