missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Decanoate 99.0+%, TCI America™
Supplier: TCI America D0024500G
| Quantity | 500 g |
|---|
Chemical Identifiers
| C10H19NaO2 | |
| MFCD00066453 | |
| sodium decanoate, sodium caprate, decanoic acid, sodium salt, decanoic acid sodium salt, sodium caprinate, capric acid sodium salt, sodium-n-decanoate, sodium decanoic acid, capric acid, sodium salt, caprinic acid sodium salt | |
| sodium decanoate |
Specifications
| Sodium Decanoate | |
| White-Yellow | |
| 500 g | |
| sodium decanoate, sodium caprate, decanoic acid, sodium salt, decanoic acid sodium salt, sodium caprinate, capric acid sodium salt, sodium-n-decanoate, sodium decanoic acid, capric acid, sodium salt, caprinic acid sodium salt | |
| [Na+].CCCCCCCCCC([O-])=O | |
| 194.25 | |
| 194.25 | |
| Crystalline Powder |
| 1002-62-6 | |
| C10H19NaO2 | |
| MFCD00066453 | |
| FIWQZURFGYXCEO-UHFFFAOYSA-M | |
| sodium decanoate | |
| 4457968 | |
| ≥99.0% (T) |
Safety and Handling
EINECSNumber : (2)-0611&(7)-0973
RTECSNumber : HE0200000
TSCA : Yes