CAS RN 1002-62-6
IUPAC Name:
sodium decanoate
Synonyms:
sodium caprate
Molecular Weight (g/mol):
194.25
Molecular Formula:
C10H19NaO2
InChi Key:
FIWQZURFGYXCEO-UHFFFAOYSA-M
SMILES:
[Na+].CCCCCCCCCC([O-])=O
1
–
1
of
1
results
1
Sodium Decanoate 99.0+%, TCI America™
CAS: 1002-62-6 Molecular Formula: C10H19NaO2 Molecular Weight (g/mol): 194.25 MDL Number: MFCD00066453 InChI Key: FIWQZURFGYXCEO-UHFFFAOYSA-M Synonym: sodium decanoate,sodium caprate,decanoic acid, sodium salt,decanoic acid sodium salt,sodium caprinate,capric acid sodium salt,sodium-n-decanoate,sodium decanoic acid,capric acid, sodium salt,caprinic acid sodium salt PubChem CID: 4457968 IUPAC Name: sodium decanoate SMILES: [Na+].CCCCCCCCCC([O-])=O