missing translation for 'onlineSavingsMsg'
Learn More

Sodium Decanoate 99.0+%, TCI America™

Catalog No. d0024100g
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0024100G 100 g
D002425G 25 g
D0024500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0024100G Supplier TCI America Supplier No. D0024100G
Only null left

Chemical Identifiers

CAS 1002-62-6
Molecular Formula C10H19NaO2
Molecular Weight (g/mol) 194.25
MDL Number MFCD00066453
InChI Key FIWQZURFGYXCEO-UHFFFAOYSA-M
Synonym sodium decanoate, sodium caprate, decanoic acid, sodium salt, decanoic acid sodium salt, sodium caprinate, capric acid sodium salt, sodium-n-decanoate, sodium decanoic acid, capric acid, sodium salt, caprinic acid sodium salt
PubChem CID 4457968
IUPAC Name sodium decanoate
SMILES [Na+].CCCCCCCCCC([O-])=O

Specifications

Color White-Yellow
Quantity 100 g
Formula Weight 194.25
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Decanoate
EINECSNumber (2)-0611&(7)-0973
RTECSNumber HE0200000
TSCA Yes