missing translation for 'onlineSavingsMsg'
Learn More

Sodium D-Pantothenate 97.0+%, TCI America™

Supplier:  TCI America P0013250G

 View more versions of this product

Catalog No. P0013250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

124°C
867-81-2
C9H16NNaO5
sodium d-pantothenate, sodium 3-2s-2,4-dihydroxy-3,3-dimethylbutanamido propanoate, natrium-d-pantothenat, ?-sodium pantothenate, ksc492i8l, unii-66y94d1203 component
[Na+].CC(C)(O)CC(O)C(=O)NCCC([O-])=O
250 g
45358008
≥97.0% (N)
Sodium D-Pantothenate
White
MFCD00002767
ROMBWECPDOCQMZ-UHFFFAOYNA-M
sodium 3-(2,4-dihydroxy-4-methylpentanamido)propanoate
241.22
241.22
Crystalline Powder

Chemical Identifiers

867-81-2
241.22
ROMBWECPDOCQMZ-UHFFFAOYNA-M
45358008
[Na+].CC(C)(O)CC(O)C(=O)NCCC([O-])=O
C9H16NNaO5
MFCD00002767
sodium d-pantothenate, sodium 3-2s-2,4-dihydroxy-3,3-dimethylbutanamido propanoate, natrium-d-pantothenat, ?-sodium pantothenate, ksc492i8l, unii-66y94d1203 component
sodium 3-(2,4-dihydroxy-4-methylpentanamido)propanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1001

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions