missing translation for 'onlineSavingsMsg'
Learn More

Sodium D-Pantothenate 97.0+%, TCI America™

Catalog No. p-7134377
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P001325G 25 g
P0013250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P001325G Supplier TCI America Supplier No. P001325G
Only null left

Chemical Identifiers

CAS 867-81-2
Molecular Formula C9H16NNaO5
Molecular Weight (g/mol) 241.22
MDL Number MFCD00002767
InChI Key ROMBWECPDOCQMZ-UHFFFAOYNA-M
Synonym sodium d-pantothenate, sodium 3-2s-2,4-dihydroxy-3,3-dimethylbutanamido propanoate, natrium-d-pantothenat, ?-sodium pantothenate, ksc492i8l, unii-66y94d1203 component
PubChem CID 45358008
IUPAC Name sodium 3-(2,4-dihydroxy-4-methylpentanamido)propanoate
SMILES [Na+].CC(C)(O)CC(O)C(=O)NCCC([O-])=O

Specifications

Melting Point 124°C
Color White
Quantity 25 g
Formula Weight 241.22
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Sodium D-Pantothenate
EINECSNumber (9)-1001
TSCA Yes
Recommended Storage Refrigerator