missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyclamate, MilliporeSigma™ Supelco™

Catalog No. 111013032
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013032 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013032 Supplier Merck Emd Millipore Supplier No. 47827
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 139-05-9
Physical Form Neat
Linear Formula C6H11 NHSO3 Na
MDL Number MFCD00003827
Melting Point >300°C
Synonym Sodium N-cyclohexylsulfamate; N-Cyclohexylsulfamic acid sodium salt; Cyclamic acid sodium salt; Cyclohexanesulfamic acid sodium salt; Sodium cyclamate; Sodium cyclohexanesulfamate
Type Food Additive Standard
Quantity 1 g
Formula Weight 201.22
Grade Analytical Standard