missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyclamate, 98%

Catalog No. p-7024286
Change view
Click to view available options
Quantity:
250 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1866630 250 g
AAA186660I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1866630 Supplier Thermo Scientific Chemicals Supplier No. A1866630

CAS: 139-05-9 | C6H12NNaO3S | 201.22 g/mol

Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose's. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Cyclamate is an artificial sweetener and is 30 times as saccharose′s. It is widely used in pickles, seasoning sauce, cakes, biscuits, bread, ice cream, frozen sucker, popsicles, drinks and so on, with a maximum amount of 0.65g/kg. Secondly, it is used in confect, with a maximum amount of 1.0g/kg. Thirdly, it is used in orange peel, preserved plum, dried arbutus and so on, with the largest amount of 8.0g/kg. It is also used in cosmetics and pharmaceutical industry.

Solubility
Soluble in water (>=10 g/100 mL at 20°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 139-05-9
Molecular Formula C6H12NNaO3S
Molecular Weight (g/mol) 201.22
MDL Number MFCD00003827
InChI Key UDIPTWFVPPPURJ-UHFFFAOYSA-M
Synonym sodium cyclamate, sodium cyclohexylsulfamate, sodium n-cyclohexylsulfamate, cyclamate sodium, cyclamic acid sodium salt, assugrin, ibiosuc, suessette, suestamin, sugarin
PubChem CID 23665706
ChEBI CHEBI:82431
SMILES [Na+].[O-]S(=O)(=O)NC1CCCCC1

Specifications

Melting Point >300°C
Quantity 250 g
Beilstein 4166868
Solubility Information Soluble in water (>=10g/100 mL at 20°C).
IUPAC Name sodium;N-cyclohexylsulfamate
Formula Weight 201.22
Percent Purity 98%
Chemical Name or Material Sodium cyclamate
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 205-348-9
RTECSNumber GV7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only