missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Cyanoborohydride 95.0+%, TCI America™
Supplier: TCI America S039625G
Specifications
| Sodium Cyanoborohydride [Reducing Agent] | |
| 25895-60-7 | |
| Crystalline Powder | |
| CH3BNNa | |
| 3134 | |
| CVDUGUOQTVTBJH-UHFFFAOYSA-N | |
| sodium cyanoboranuide | |
| 20587905 | |
| ≥95.0% (T) |
| White-Yellow | |
| 8 to 9 (10% H2O soln.) | |
| 25 g | |
| MFCD00003516 | |
| sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1- | |
| [Na+].[BH3-]C#N | |
| 62.84 | |
| 62.84 |
Chemical Identifiers
| 25895-60-7 | |
| 62.84 | |
| CVDUGUOQTVTBJH-UHFFFAOYSA-N | |
| 20587905 | |
| [Na+].[BH3-]C#N |
| CH3BNNa | |
| MFCD00003516 | |
| sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1- | |
| sodium cyanoboranuide |
Safety and Handling
TSCA : Yes