missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanoborohydride 95.0+%, TCI America™

Catalog No. p-7135878
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03965G 5 g
S039625G 25 g
S0396250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S03965G Supplier TCI America Supplier No. S03965G
Only null left

Chemical Identifiers

CAS 25895-60-7
Molecular Formula CH3BNNa
Molecular Weight (g/mol) 62.84
MDL Number MFCD00003516
InChI Key CVDUGUOQTVTBJH-UHFFFAOYSA-N
Synonym sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
PubChem CID 20587905
IUPAC Name sodium cyanoboranuide
SMILES [Na+].[BH3-]C#N

Specifications

Color White-Yellow
pH 8 to 9 (10% H2O soln.)
Physical Form Crystalline Powder
Quantity 5 g
UN Number 3134
Formula Weight 62.84
Percent Purity ≥95.0% (T)
Chemical Name or Material Sodium Cyanoborohydride [Reducing Agent]