missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Cyanide, ACS Reagent, ≥95.0%, Honeywell Fluka™
ACS Reagent,≥95.0%
$1417.39 - $1417.39
Chemical Identifiers
| CAS | 143-33-9 |
|---|---|
| Molecular Formula | CNNa |
| Molecular Weight (g/mol) | 49.01 |
| MDL Number | MFCD00003523 |
| InChI Key | MNWBNISUBARLIT-UHFFFAOYSA-N |
| Synonym | sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h |
| PubChem CID | 8929 |
| IUPAC Name | sodium;cyanide |
| SMILES | [Na+].[C-]#N |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 143-33-9 | |
| 49.01 | |
| MNWBNISUBARLIT-UHFFFAOYSA-N | |
| 8929 | |
| [Na+].[C-]#N |
| CNNa | |
| MFCD00003523 | |
| sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h | |
| sodium;cyanide |
Specifications
| 1407°C | |
| 143-33-9 | |
| 11 to 12 (49g/L at 25°C) | |
| CNNa | |
| MFCD00003523 | |
| 3587243 | |
| Soluble in water | |
| [Na+].[C-]#N | |
| 49.01 | |
| 49.01g/mol | |
| ACS Reagent | |
| Crystals | |
| 1.6g/cu.cm; Vapor Density: 1.7 (vs air) |
| 563°C | |
| Colorless | |
| 2.5 kg | |
| NaCN | |
| UN1689 | |
| sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h | |
| MNWBNISUBARLIT-UHFFFAOYSA-N | |
| sodium;cyanide | |
| 8929 | |
| ≥95.0% | |
| Poly Bottle | |
| 1mmHg (817°C) | |
| Sodium Cyanide |
Safety and Handling
P260-P273-P280-P284-P302 + P352 + P310-P304 + P340 + P310
H290-H300 + H310 + H330-H372-H410
DOTInformation : 6.1/PGI
EINECSNumber : 205-599-4
RTECSNumber : VZ7525000