missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, ACS Reagent, ≥95.0%, Solstice

Catalog No. 60045111
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045111 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60045111 Supplier Honeywell Inc Supplier No. 2052222.5KG

missing translation for 'validMainframeMsg'

ACS Reagent,≥95.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
IUPAC Name sodium;cyanide
SMILES [Na+].[C-]#N

Specifications

Boiling Point 1407°C
Melting Point 563°C
Color Colorless
pH 11 to 12 (49g/L at 25°C)
Physical Form Crystalline Powder
Quantity 2.5 kg
Linear Formula NaCN
UN Number UN1689
Beilstein 3587243
Solubility Information Soluble in water
Formula Weight 49.01g/mol
Percent Purity ≥95.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystals
Vapor Pressure 1mmHg (817°C)
Density 1.6g/cu.cm; Vapor Density: 1.7 (vs air)
Chemical Name or Material Sodium Cyanide
Show More Show Less
Hazard Statement P260-P273-P280-P284-P302 + P352 + P310-P304 + P340 + P310
Precautionary Statement H290-H300 + H310 + H330-H372-H410
DOTInformation 6.1/PGI
EINECSNumber 205-599-4
RTECSNumber VZ7525000