missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cobaltinitrite, Powder, Reagent, ACS, Spectrum™ Chemical

Catalog No. p-7158208
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606466 25 g Amber Glass Bottle
18606468 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606466 Supplier Spectrum Chemical Mfg Cor Supplier No. S125525GM
Only null left

S1255, 13600-98-1, Na3Co(NO2)6

Spectrum™ Chemical Sodium Cobaltinitrite, Powder, Reagent, ACS , a coordination compound, forms the basis of a quantitative determination of thallium, potassium, and ammonium ions. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 13600-98-1
Molecular Formula CoN6Na3O12
Molecular Weight (g/mol) 403.93
MDL Number MFCD00003512
InChI Key BZFWEBJDBATXME-UHFFFAOYSA-N
IUPAC Name trisodium hexanitrocobalttris(ylium)
SMILES [Na+].[Na+].[Na+].[O-][N+](=O)[Co+3]([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Max % 1
Quantity 25 g
Grade ACS
Packaging Amber Glass Bottle