missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Sodium Citrate USP/EP (Dihydrate)
Description
Sodium Citrate USP / EP is a white or almost white, crystalline powder or white or almost white, granular crystals. It is slightly deliquescent in moist air.
Alternative name(s): sodium citrate dihydrate, trisodium citrate dihydrate, trisodium citrate
Specifications
Specifications
| Chemical Name or Material | Sodium Citrate USP/EP (Dihydrate) |
| Packaging | Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck |
| CAS | 6132-04-3 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 294.1 |
| Quantity | 2.5 kg |
| Grade | EP, USP |