missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium citrate, 0.5M buffer soln., pH 5.5

Catalog No. AAJ63199AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63199AP 500 mL
AAJ63199AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63199AP Supplier Thermo Scientific Chemicals Supplier No. J63199AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium citrate is commonly used in nucleic acid hybridization techniques at concentrations ranging from 0.1X to 20X depending on the application.  It is used to facilitate transfer of nucleic acids to membranes in southern and northern blotting, dot blotting, and colony and plaque lifts. 

Solubility
Soluble in water.

Notes
20X SSC Solution (RNase-free) should be stored at room temperature. This reagent should remain stable for at least 1 year in its unopened container.

Specifications

Chemical Name or Material Sodium citrate buffer soln.
Color Colorless
Concentration 0.5 M
pH 5.5
Physical Form Liquid
Quantity 500 mL
Odor Odorless
MDL Number MFCD00012462
Solubility Information Soluble in water.

RUO – Research Use Only