missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Cholate, Reagent, 95%, Spectrum™ Chemical
SO162, 361-09-1, C24H39NaO5
$192.53 - $112059.33
Chemical Identifiers
| CAS | 361-09-1 |
|---|---|
| Molecular Formula | C24H39NaO5 |
| Molecular Weight (g/mol) | 430.56 |
| MDL Number | MFCD00150749,MFCD00064138 |
| InChI Key | NRHMKIHPTBHXPF-TUJRSCDTSA-M |
| IUPAC Name | sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate |
| SMILES | [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606437
|
Spectrum Chemical Mfg Cor
SO1625GM |
5 g | Amber Glass Bottle |
Each for $192.53
|
|
||||
|
18606438
|
Spectrum Chemical Mfg Cor
SO16225GM |
25 g | Poly Bottle |
Each for $340.33
|
|
||||
|
18606439
|
Spectrum Chemical Mfg Cor
SO162100GM |
100 g | Poly Bottle |
Each for $684.83
|
|
||||
|
18606440
|
Spectrum Chemical Mfg Cor
SO162500GM |
500 g | Poly Bottle |
Each for $2,394.83
|
|
||||
|
18606441
|
Spectrum Chemical Mfg Cor
SO1621KG |
1 kg | Poly Pail |
Each for $4,025.90
|
|
||||
|
18606442
|
Spectrum Chemical Mfg Cor
SO1625KG |
5 kg | Poly Pail |
Each for $9,995.20
|
|
||||
|
18606443
|
Spectrum Chemical Mfg Cor
SO16250KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sodium Cholate, Reagent The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
| 361-09-1 | |
| 430.56 | |
| NRHMKIHPTBHXPF-TUJRSCDTSA-M | |
| [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C |
| C24H39NaO5 | |
| MFCD00150749,MFCD00064138 | |
| sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate |
Specifications
| 361-09-1 | |
| 5 g | |
| MFCD00150749,MFCD00064138 | |
| [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C | |
| 430.56 | |
| Reagent |
| 1 | |
| C24H39NaO5 | |
| NRHMKIHPTBHXPF-TUJRSCDTSA-M | |
| sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate | |
| 95% | |
| Amber Glass Bottle |