accessibility menu, dialog, popup

UserName

Explore New Spotlight Deals on Equipment, Instruments, and More

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 361-09-1

NaCH3(R)OO(R)(S)(R)(R)OH(S)(R)HO(S)CH3(S)(S)OH(R)CH3

CAS RN 361-09-1

IUPAC Name: sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
Synonyms: sodium cholate cholic acid, monosodium salt
Molecular Weight (g/mol): 430.56
Molecular Formula: C24H39NaO5
InChi Key: NRHMKIHPTBHXPF-TUJRSCDTSA-M
SMILES: [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
Percent Purity 
  • (1)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (7)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.