missing translation for 'onlineSavingsMsg'
Learn More

Sodium cholate hydrate, 99%, Thermo Scientific Chemicals

Catalog No. AAA1707418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1707418 50 g
AAA1707409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1707418 Supplier Thermo Scientific Chemicals Supplier No. A1707418

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biochemical solubilizing agent Sodium cholate hydrate is a non-denaturing detergent used for the extraction of membrane proteins. It is involved in the study to assess infiltration of pressurized promoter solutions in a mesoporous silica. Further, it serves as a biochemical solubilizing agent.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong acids.

Chemical Identifiers

CAS 206986-87-0
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.56
MDL Number MFCD00064138 MFCD00003672
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
PubChem CID 23679061
IUPAC Name sodium;(4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate;hydrate
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC([O-])=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Quantity 50 g
Beilstein 3582354
Synonym sodium cholate hydrate, cholalic acid sodium salt, c24h39o5.na.h2o, sodium cholate hydrate, bioxtra, sodium cholate hydrate dried material, nt, sodium cholate hydrate, from ox and/or sheep bile, 3, a,7, a,12, a-trihydroxy-5, a-cholan-24-oic acid sodium salt, sodium cholate hydrate, suitable for cell culture, bioreagent, 4r-4-1s,2s,7s,11s,16s,5r,9r,10r,14r,15r-5,9,16-trihydroxy-2,15-dimethylte tracyclo 8.7.0.0<2,7>.0<11,15> heptadec-14-yl pentanoic acid, sodium salt, hyd rate, sodium cholate hydrate, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
Solubility Information Soluble in water.
Optical Rotation +32° (c=0.5 in Water)
Formula Weight 430.56 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Sodium cholate hydrate
RTECSNumber FZ9600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only