missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorodifluoroacetate 99.0+%, TCI America™

Catalog No. p-7124729
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
C099125G 25 g
C0991100G 100 g
C0991500G 500 g
3 options

Catalog No. C099125G

Supplier: TCI America C099125G

Only null left

Chemical Identifiers

CAS 1895-39-2
Molecular Formula C2ClF2NaO2
Molecular Weight (g/mol) 152.457
MDL Number MFCD00064771
InChI Key MRTAVLDNYYEJHK-UHFFFAOYSA-M
Synonym sodium chlorodifluoroacetate, sodium 2-chloro-2,2-difluoroacetate, chlorodifluoroacetic acid sodium salt, sodium chloro difluoro acetate, acetic acid, chlorodifluoro-, sodium salt, sodiumchlorodifluoroacetate, difluorochloroacetic acid sodium salt, sodium difluorochloroacetate, chlorodifluoroacetic acid sodium
PubChem CID 2734985
IUPAC Name sodium;2-chloro-2,2-difluoroacetate
SMILES C(=O)(C(F)(F)Cl)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 152.46
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Chlorodifluoroacetate