Learn More
Sodium chloroacetate, 98%
CAS: 3926-62-3 | C2H2ClNaO2 | 116.476 g/mol
Supplier: Thermo Scientific Chemicals A123790E
Description
Sodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium chloroacetate is used to prepare dyes and active pharmaceutical ingredients. It is also used as an odor agent, surface active agent and viscosity adjustor.
Solubility
Soluble in water, ether, chloroform, benzene and alcohol.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Sodium chloroacetate | |
| ∼199°C (decomposition) | |
| 270°C (518°F) | |
| Characteristic | |
| ClCH2CO2Na | |
| 2500 g | |
| 3597157 | |
| sodium chloroacetate, chloroacetic acid sodium salt, monoxone, acetic acid, chloro-, sodium salt, dow defoliant, sodium monochloroacetate, smca, sodium monochloracetate, caswell no. 755a, chloroctan sodny czech | |
| FDRCDNZGSXJAFP-UHFFFAOYSA-M | |
| sodium;2-chloroacetate | |
| 23665759 | |
| 98% |
| 3926-62-3 | |
| 6 | |
| 98% | |
| C2H2ClNaO2 | |
| MFCD00002684 | |
| UN2659 | |
| 14,2112 | |
| Soluble in water,ether,chloroform,benzene and alcohol. | |
| C(C(=O)[O-])Cl.[Na+] | |
| 116.476 | |
| 116.48 | |
| Powder |
Chemical Identifiers
| 3926-62-3 | |
| 116.476 | |
| FDRCDNZGSXJAFP-UHFFFAOYSA-M | |
| 23665759 | |
| C(C(=O)[O-])Cl.[Na+] |
| C2H2ClNaO2 | |
| MFCD00002684 | |
| sodium chloroacetate, chloroacetic acid sodium salt, monoxone, acetic acid, chloro-, sodium salt, dow defoliant, sodium monochloroacetate, smca, sodium monochloracetate, caswell no. 755a, chloroctan sodny czech | |
| sodium;2-chloroacetate |
Safety and Handling
GHS H Statement
H301-H315
Toxic if swallowed.
Causes skin irritation.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H301-H315
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SODIUM CHLOROACETATE
EINECSNumber : 223-498-3
RTECSNumber : AG1400000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only