missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorite, Anhydrous, Flakes, 77.5-82.5%, Spectrum™ Chemical

Catalog No. p-7158202
Change view
Click to view available options
Quantity:
100 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606432 100 g Amber Glass Bottle
18606434 2.5 kg Poly Pail
18606435 12 kg Poly Pail
18606436 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606432 Supplier Spectrum Chemical Mfg Cor Supplier No. S1243100GM
Only null left

S1243, 7758-19-2, NaClO2

Spectrum™ Chemical Sodium Chlorite, Anhydrous, Flakes is a chemical compound often used in the manufacture of paper. Used to generate chlorine dioxide for the bleaching and striping of textiles it is also frequently used as a oxidation reagent in organic synthesis.

(CAS No. 7758-19-2, Molecular Formula: NaClO2) Sodium Chlorite, Anhydrous, Flakes

Chemical Identifiers

CAS 7758-19-2
Molecular Formula ClNaO2
Molecular Weight (g/mol) 90.44
InChI Key UKLNMMHNWFDKNT-UHFFFAOYSA-M
IUPAC Name sodium chlorite
SMILES [Na+].[O-][Cl]=O

Specifications

CAS Max % 1
Quantity 100 g
Percent Purity 77.5 to 82.5%
Grade Reagent
Packaging Amber Glass Bottle