missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorite, Anhydrous, Flakes, 77.5-82.5%, Spectrum™ Chemical

Catalog No. 18606432
Change view
Click to view available options
Quantity:
100 g
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606432 100 g Amber Glass Bottle
18606435 12 kg Poly Pail
18606436 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606432 Supplier Spectrum Chemical Mfg Cor Supplier No. S1243100GM

S1243, 7758-19-2, NaClO2

Spectrum™ Chemical Sodium Chlorite, Anhydrous, Flakes is a chemical compound often used in the manufacture of paper. Used to generate chlorine dioxide for the bleaching and striping of textiles it is also frequently used as a oxidation reagent in organic synthesis.

(CAS No. 7758-19-2, Molecular Formula: NaClO2) Sodium Chlorite, Anhydrous, Flakes

Chemical Identifiers

CAS 7758-19-2
Molecular Formula ClNaO2
Molecular Weight (g/mol) 90.44
InChI Key UKLNMMHNWFDKNT-UHFFFAOYSA-M
IUPAC Name sodium chlorite
SMILES [Na+].[O-][Cl]=O

Specifications

CAS Max % 1
Quantity 100 g
Percent Purity 77.5 to 82.5%
Grade Reagent
Packaging Amber Glass Bottle