Learn More
Sodium chlorite, 80%, pure, unstabilized
Sodium chlorite, 80%, ClNaO2, CAS Number-7758-19-2, 7647-14-5, unii-g538ebv4vf, alcide ld, neo silox d, chlorous acid, sodium salt, caswell no. 755, sodiumchlorite, textone, sodium chlorite, textile, chlorite sodium, 500g, CHEBI:78667, White, 231-836-6, 05,603; 09,423; 11,481; 13,280, 90.44, Pure | CAS: 7758-19-2 | ClNaO2 | 90.44 g/mol
Supplier: Thermo Scientific Chemicals 223230025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium chlorite | |
| Unstabilized | |
| White | |
| 2.5 kg | |
| ClNaO2 | |
| MFCD00003478 | |
| 15, 8735 | |
| Solubility in water: 572g/L (20°C) | |
| [O-]Cl=O.[Na+] | |
| 90.44 | |
| CHEBI:78667 | |
| 80% | |
| Plastic Bottle |
| 180.0°C to 200.0°C | |
| 7758-19-2,7647-14-5 | |
| Powder | |
| 75% min. (Iodometry) | |
| NaClO2 | |
| 05,603; 09,423; 11,481; 13,280 | |
| sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf | |
| UKLNMMHNWFDKNT-UHFFFAOYSA-M | |
| sodium;chlorite | |
| 23668197 | |
| 90.44 | |
| Pure |
Chemical Identifiers
| 7758-19-2 | |
| UKLNMMHNWFDKNT-UHFFFAOYSA-M | |
| 23668197 | |
| sodium;chlorite |
| MFCD00003478 | |
| sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf | |
| CHEBI:78667 | |
| [O-]Cl=O.[Na+] |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cauti
GHS Signal Word: Danger
EINECSNumber : 231-836-6
RTECSNumber : VZ4800000
TSCA : TSCA
RUO – Research Use Only