missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, USP, EP, JP, FCC, Pyrogen Free, 99-100.5%, Macco Organiques

Catalog No. p-7184408
Change view
Click to view available options
Quantity:
12 kg
50 kg
Packaging:
Plastic Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614845 12 kg Plastic Pail
18615333 50 kg Plastic Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614845 Supplier Spectrum Chemical Mfg Cor Supplier No. 51103000120
Only null left

Macco products offered by Spectrum™ Chemical are suitable for biopharmaceutical production.

  • Full testing including bioburden, endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color White or Almost White
Physical Form Crystalline Powder
Quantity 12 kg
Solubility Information Freely soluble in water, practically insoluble in anhydrous ethanol
Percent Purity 99 to 100.5%
Grade EP/FCC/JP/USP
Loss on Drying 0.5%
Packaging Plastic Pail