missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, USP, EP, 99-100.5%, Spectrum™ Chemical
S1529, 7647-14-5, NaCl
$1481.04 - $2275.71
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |