missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Chloride Tablets, Calbiochem™,

Catalog No. 5674421EA
Change view
Click to view available options
Quantity:
10 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5674421EA 10 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5674421EA Supplier MilliporeSigma™ Supplier No. 5674421EA

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym NaCl 10 Tablets
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Quantity 10 Ea.
Packaging Glass Bottle