missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, Spectroscopy Grade

Catalog No. AA4363119
Change view
Click to view available options
Quantity:
75 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4363119 75 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4363119 Supplier Thermo Scientific Chemicals Supplier No. 04363119

CAS: 7647-14-5 | ClNa | 58.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1413°C
Melting Point 801°C
Color White
pH 6 to 9
Physical Form Powder
Quantity 75 g
Merck Index 14,8599
Formula Weight 58.44
Grade Spectroscopy
Odor Odorless
Refractive Index 1.5442
Sensitivity Hygroscopic
Density 2.165 g/mL
Chemical Name or Material Sodium chloride
Show More Show Less
EINECSNumber 231-598-3
RTECSNumber VZ4725000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only