missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, ReagentPlus™, Honeywell™

Catalog No. p-7072320
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
10 kg
5 kg
Catalog No. Quantity
6003190 1 kg
6003192 5 kg
6003189 10 kg
3 options

Catalog No. 6003190

Supplier: Honeywell Chemicals S96251KG

Only null left

Reagent Grade,≥99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Quantity 1 kg
Linear Formula NaCl
UN Number NONH for all modes of transport
Beilstein 3534976
IUPAC Name sodium;chloride
Formula Weight 58.44g/mol
Percent Purity ≥99%
Grade ReagentPlus
Chemical Name or Material Sodium chloride
RTECSNumber VZ4725000