missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, puriss. p.a., Honeywell Fluka™
Puriss. p.a.,≥99.5% (AT)
$282.82 - $916.71
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| SMILES | [Na+].[Cl-] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 |
Specifications
| 7647-14-5 | |
| ClNa | |
| MFCD00003477 | |
| 3534976 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 58.44g/mol | |
| puriss. p.a. |
| 1 kg | |
| NaCl | |
| NONH for all modes of transport | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| ≥99.5% (AT) | |
| Sodium chloride |
Safety and Handling
RTECSNumber : VZ4725000