missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al, Honeywell Fluka™
Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al
Supplier: Honeywell-Fluka 134235KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Sodium Chloride | |
| 7647-14-5 | |
| ClNa | |
| MFCD00003477 | |
| 3534976 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 58.44g/mol | |
| puriss. |
| Meets analytical specification of Ph. Eur.,BP, USP | |
| 5 kg | |
| NaCl | |
| NONH for all modes of transport | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| 99 to 100.5% (calc. to the dried substance) |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| [Na+].[Cl-] |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 |
Safety and Handling
EINECSNumber : 231-598-3
RTECSNumber : VZ4725000