missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Purified, Reagents

Catalog No. p-200021090
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
50 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337040 10 kg
01337039 2.5 kg
01337038 500 g
01337037 50 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337040 Supplier Labchem Inc Supplier No. C231300010KG
Only null left

Sodium Chloride, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym Table salt
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color Colorless/White
pH 6.7 to 7.3
Physical Form Solid
Quantity 10 kg
Grade Pure
Packaging Poly Pail
Specific Gravity 2.17
Chemical Name or Material Sodium Chloride, Purified