missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015463
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015463 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015463 Supplier Merck Emd Millipore Supplier No. PHR13215G
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Sodium chloride is an ionic compound used mainly as a preservative and flavor forfood. Apart from this, it is widely used in industries for manufacturing different products such as paper, soaps, detergents, rubber, dyes and glass etc. It occurs naturally in soils and aids in diverse cellular functions within the human body.

Specifications

CAS 7647-14-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Linear Formula NaCl
MDL Number MFCD00003477
Melting Point 801°C
Synonym Halite
Type Certified Reference Material
Quantity 5 g
Formula Weight 58.44
Grade Certified Reference Material