missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, Optical Grade

CAS: 7647-14-5 | ClNa | 58.44 g/mol

$204.78 - $778.49

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1101918
View Documents
Thermo Scientific Chemicals
01101918
50 g
Each for $204.78
Only null left
 
AA1101930
View Documents
Thermo Scientific Chemicals
01101930
250 g
Each for $778.49
Only null left
 
Description

Description

Sodium Chloride (NaCl) is widely used in biochemistry and molecular biology applications. It is considered an essential nutrient. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. NaCl is a component of phosphate buffered saline and SSC buffer. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]

Specifications

1413°C
7647-14-5
6 to 9
50 g
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
58.44
Odorless
Hygroscopic
Sodium chloride
801°C
White
Broken Crystals
ClNa
14,8599
Soluble in water at 20°C 358g/L. Very slightly soluble in alcohol,glycerol (100mg/ml),and ammonia. Insoluble in hydrochloric acid.
[Na+].[Cl-]
58.44
CHEBI:26710
Optical
1.5442
2.165 g/mL
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-598-3

RTECSNumber : VZ4725000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only