missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Sodium Chloride, OmniPur™, Calbiochem™,
$89.22 - $703.24
Chemical Identifiers
| CAS | 7647-14-5 |
|---|---|
| Molecular Formula | ClNa |
| Molecular Weight (g/mol) | 58.44 |
| MDL Number | MFCD00003477 |
| InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
| Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
| PubChem CID | 5234 |
| ChEBI | CHEBI:26710 |
| IUPAC Name | sodium chloride |
| SMILES | [Na+].[Cl-] |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
Specifications
| 1430.8°C | |
| 500 g | |
| ClNa | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 58.44 | |
| Sodium Chloride |
| 7647-14-5 | |
| ≥99.5% | |
| MFCD00003477 | |
| Soluble in water | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| Odorless |
Safety and Handling
Recommended Storage : Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.