missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Chloride, OmniPur™, Calbiochem™,

Catalog No. 7710500GM
Change view
Click to view available options
Quantity:
500 g
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7710500GM 500 g
778025KG 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7710500GM Supplier MilliporeSigma™ Supplier No. 7710500GM

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1430.8°C
Quantity 500 g
Assay Percent Range ≥99.5%
Solubility Information Soluble in water
Formula Weight 58.44
Odor Odorless
Chemical Name or Material Sodium Chloride
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.