missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Fisher BioReagents™

Catalog No. BP358212
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP358212 2.5 kg Poly Bottle
BP3581 1 kg Poly Bottle
BP35810 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP358212 Supplier Fisher BioReagents Supplier No. BP358 212
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Sodium Chloride

Sodium Chloride is a main component of many electrophoresis and molecular biology buffers and media.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 2.5 kg
Assay Percent Range ≥99.0 %
Merck Index 15, 8734
IUPAC Name sodium chloride
Formula Weight 58.44
Percent Purity ≥99.0%
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 1mmHg at 440°C
Density 2.165g/cm³
Assay (Dry Basis) >99.5%
Chemical Name or Material Sodium Chloride
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1
Recommended Storage RT