missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, extra pure

CAS: 7647-14-5 | ClNa | 58.44 g/mol

$108.70 - $300.83

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC194090010
SDS View Documents
Thermo Scientific Chemicals
194090010
1 kg Plastic Bottle
Each for $108.70
Only null left
 
AC194090050
SDS View Documents
Thermo Scientific Chemicals
194090050
5 kg Plastic Bottle
Each for $300.83
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
[Na+].[Cl-]
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710

Specifications

1461.0°C
7647-14-5
5.0 to 8.0 (5%, H<2O, at 20°C)
1 kg
NaCl
13,280
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
58.44
Extra Pure
Sodium chloride
801.0°C
White
Crystalline Powder
ClNa
MFCD00003477
15, 8734
Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid
[Na+].[Cl-]
58.44
CHEBI:26710
≥99.0%
Plastic Bottle
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-598-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only