missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, crystalline powder PDV, 99+%

Catalog No. p-7082058
Change view
Click to view available options
Quantity:
25,000 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA123130B 1000 g
AAA123130I 5000 g
AAA123130G 25,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA123130B Supplier Thermo Scientific Chemicals Supplier No. A123130B
Only null left

CAS: 7647-14-5 | ClNa | 58.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium chloride is widely used as a food additive, and a preservative. It is employed in the Chloralkali process for producing chlorine and sodium hydroxide, in the Solvay process for producing sodium carbonate and calcium chloride, in the Mannheim process and in the Hargreaves process for producing sodium sulfate and hydrochloric acid. It is used in the manufacture of sodium chlorate, water softening and de-icing of roadways. Further it is used in several industries like pharmaceutical, textile, dyeing, rubber food and agriculture. It is used in the processing of steel, copper, vanadium, and other metals.

Solubility
Freely soluble in water. Slightly soluble in alcohol and ammonia.

Notes
It is sensitive to moisture in air. It should be protected in an air tight container.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1413°C
Melting Point 801°C
pH 6 to 9
Physical Form Crystalline Powder
Quantity 1000 g
Merck Index 14,8599
Solubility Information Freely soluble in water. Slightly soluble in alcohol and ammonia.
Formula Weight 58.44
Percent Purity ≥99%
Odor Odorless
Refractive Index 1.5442
Sensitivity Hygroscopic
Density 2.165
Chemical Name or Material Sodium chloride
Show More Show Less
EINECSNumber 231-598-3
RTECSNumber VZ4725000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only