missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm, unpolished, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 38397KT

Quantity 1 ea.

Catalog No. AA38397KT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium;chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]

Specifications

Sodium chloride crystal optic rectangle
7647-14-5
ClNa
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
[Na+].[Cl-]
58.44
CHEBI:26710
38.5mm x 19.5mm x 4mm, Unpolished
1 ea.
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium;chloride
5234
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-598-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents