missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride, Certified, 25% (w/v) ±1%, LabChem™
Supplier: LabChem LC235204
Specifications
| Sodium Chloride | |
| Colorless | |
| Liquid | |
| ClNa | |
| MFCD00003477 | |
| Soluble in water | |
| [Na+].[Cl-] | |
| 58.44 | |
| CHEBI:26710 | |
| 25% ±1% (w/v) | |
| Passes Test | |
| 1.15g/mL | |
| 1.15g/mL |
| 7647-14-5,7732-18-5 | |
| 79.21,20.79 | |
| 4 L | |
| NaCl | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| sodium chloride | |
| 5234 | |
| 58.44 | |
| Certified | |
| Poly Bottle | |
| Hydrogen chloride |
Chemical Identifiers
| 7647-14-5 | |
| 58.44 | |
| FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
| 5234 | |
| sodium chloride |
| ClNa | |
| MFCD00003477 | |
| sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
| CHEBI:26710 | |
| [Na+].[Cl-] |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature